Learn More
Invitrogen™ ZOOM™ CHAPS
For use with ZOOM IEF Fractionator
Brand: Invitrogen™ ZC10003
171.50 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Description
ZOOM CHAPS ensure optimal conditions for sample preparation.
IsoElectric Focusing, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
The ZOOM™ IEF Fractionator Combo Kit includes the ZOOM™ IEF Fractionator; one pack ZOOM™ Disks of each pH; and ZOOM™ Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers). ZOOM™ Disks, Carrier Ampholytes, and Buffers are stored at +4°C. | |
Room Temperature | |
Zoom Reagents |
Powder | |
ZOOM™ | |
5 g |
For Research Use Only. Not for use in diagnostic procedures.