Learn More
Invitrogen™ Novex™ Zymogram Renaturing Buffer (10X)
Used with Novex™ Zymogram Gels
Brand: Invitrogen™ LC2670
Code : 50
Additional Details : Weight : 0.36000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature | |
10 X | |
Zymogram Renaturing Buffer | |
Novex™ |
Renaturing Buffers | |
Zymogram | |
Novex™ Zymogram Gels | |
Room Temperature |
Safety and Handling
- ZYMOGRAM RENAT BUF(10X)500ML
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Triton X-100
For Research Use Only. Not for use in diagnostic procedures.