Learn More
Novus Biologicals™ Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP1-06640PEP
Additional Details : Weight : 0.00970kg
Description
A peptide to KDM6B. The Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
23135 | |
Store at -80 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-06640. For Further Blocking Peptide Related Protocol, Click Here . | |
KDM6B | |
0.05 mg |
Human | |
Peptide dissolved in dH2O. Contains no BSA. | |
EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6 B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6 B, Lysine demethylase 6 B | |
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
RUO - research use only |