missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Normal Goat Serum
This product has been tested by SDS-PAGE and is ideal for blocking procedures such as Western Blotting, ELISA and immunochemistry to prevent nonspecific binding. Use in Immunohistochemistry-Paraffin reported in scientific literature (PMID: 26004262)
Brand: Novus Biologicals™ NBP2-23475
This item is not returnable.
View return policy
Description
- Lyophilized white powder or clear liquid.
- Goat Serum blocking reagent/NGS was prepared from normal serum by multi-step process which includes delipidation, selective precipitation
- Buffer: 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH: 7.2 and 0.01% (w/v) Gentamicin Sulfate
- 0.01% Sodium Azide as preservative
- Restore with deionized water (or equivalent)
Applications:
Western Blot, ELISA, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Recommended Storage
Store at 4°C for short term and -20°C for long term
Specifications
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
Multi-step Purification | |
0.01% Sodium Azide, 0.01% Gentamicin Sulfate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction