Learn More
Novus Biologicals™ ROR alpha Plasmid
For expression of hROR alpha protein in mammalian cells, induction of G6Pase promoter activity
Brand: Novus Biologicals™ NBP2-31076
Additional Details : Weight : 0.00970kg
Description
The ROR alpha Plasmidis a mammalian expression vector construct, pCMV/hRORalpha, containing coding sequence of human RAR-related orphan receptor alpha.
- Species: Human
- Immunogen: ROR alpha Plasmid Expression of hROR alpha protein in mammalian cells. Induction of G6Pase promoter activity
- Total contents of 10μg is enough for one 96-well cell transfection
Recommended Storage
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Specifications
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
ROR alpha Plasmid | |
10 μg | |
hROR, hROR alpha ROR, ROR alpha |