missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tuttnauer Nozzle Spring
Spring to adjust the nozzle height to suit different types of glassware.
£6.70 - £14.30
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17878219
|
Tuttnauer
D00543700 |
Nozzles (2.5 mm), Gasket Rack 2 CASS. PIP |
£14.30
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17848229
|
Tuttnauer
D00542200 |
Nozzles (4 mm), Stopper SIL (5 mm H) |
£6.70
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17858219
|
Tuttnauer
D00540700 |
Nozzles (6 mm) |
£6.70
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Spring to adjust the Nozzle height to suit different types of glassware.
Specifications
Stainless Steel | |
Nozzle Spring |