missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ NP-40 lysis buffer, high salt
£30.80 - £44.40
Specifications
Color | Colorless |
---|---|
Content And Storage | Keep cold |
Format | Viscous Liquid |
Formulation | 50mM Tris-HCl (pH 7.4), 500mM NaCl, 1% NP-40 and 5mM EDTA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15404039
|
Thermo Scientific™
J61428.AK |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15414039
|
Thermo Scientific™
J61428.AP |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Colorless | |
Keep cold | |
Viscous Liquid | |
50mM Tris-HCl (pH 7.4), 500mM NaCl, 1% NP-40 and 5mM EDTA |