missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NP-40 permeating solution in PBS (10X)

£31.56 - £45.41

Specifications

Color Colorless
Concentration 10X
Content And Storage Ambient temperatures
Format Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493539
View Documents
Thermo Scientific™
J60838.AK
250 mL
£31.56
250mL
Please to purchase this item. Need a web account? Register with us today!
15403549
View Documents
Thermo Scientific™
J60838.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Colorless
10X
Ambient temperatures
Liquid
Videos
SDS
Documents

Documents

Product Certifications