Learn More
Masterflex™ NPT Male Adapter to Hose Barb
Quick and easy assembly
Brand:  Masterflex™ 40610-38
Code : A0
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 13457426
  • £28.80 / Pack of 10
Estimated Shipment: 25-10-2024
to see stock.
Add to basket
Description
Description
  • Available in a variety of materials and configurations.
  • Adapts from a NPT male thread to a hose barb.
Specifications
Specifications
NPT Male Adapter to Hose Barb, 1/4 in. NPT to 1/4 in. ID
Polypropylene
1/4 in. NPT to 1/4 in. ID
Barb x NPT(M)
10/Pk.
Ethylene Oxide, Gamma