missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1H2 Mouse anti-Human, Porcine, Clone: 1E12B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60345-1-IG-20UL
This item is not returnable.
View return policy
Description
LXR-B belongs to the Liver X Receptor family that encodes highly homologous transcription factors. Like the highly homologous LXR-A, LXR-B forms heterodimers with the retinoic acid receptor RXRalpha, which function as sensors for cellular oxysterols which when activated, increase the expression of genes that control sterol and fatty acid metabolism and homeostasis. Recent experiments have indicated that the LXRs can also modulate both innate and adaptive immune responses. Other studies suggest that genetic variability at the LXR-B gene locus may be a risk factor for Alzheimer′s disease. One hypothesis postulates that LXR may be upstream of ApoE and potentiates the risk associated effects of the epsilon3 allele.Specifications
NR1H2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55055 | |
NR1H2 | |
NR1H2 Fusion Protein Ag5635 | |
20 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Western Blot | |
1E12B10 | |
Unconjugated | |
NR1H2 | |
Liver X receptor beta, LXR b, LXRB, NER, NER I, NR1H2, Nuclear receptor NER, Oxysterols receptor LXR beta, RIP15, UNR | |
Mouse | |
Protein G | |
RUO | |
100525290, 7376 | |
-20°C | |
Liquid |