missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NS 309, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive modulator of small- and intermediate- conductance Ca2+-activated K+ channels (KCa2 and KCa3.1 channels); increases Ca2+ sensitivity. Displays no activity at BK channels.
Specifications
Quantity | 10 mg |
Formula Weight | 231.04 |
Percent Purity | >98% |
Chemical Name or Material | NS 309 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'