missing translation for 'onlineSavingsMsg'
Learn More

NSC319726, MedChemExpress

Product Code. p-200101974
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460028

Brand: Medchem Express HY18634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC319726 (ZMC1) is a mutant p53R175 reactivator; inhibits growth of fibroblasts expressing the p53R175 mutation (IC50 = 8 nM); shows no inhibition for p53 wild-type cells.

NSC319726 (ZMC1) is a mutant p53R175 reactivator

Specifications

Chemical Name or Material NSC319726
CAS 71555-25-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14N4S
Quantity 5 mg
Synonym ZMC1
Solubility Information DMSO : 31.25 mg/mL (133.36 mM; Need ultrasonic)
SMILES C/C(C1=NC=CC=C1)=N\NC(N2CCC2)=S
Molecular Weight (g/mol) 234.32
Formula Weight 234.32
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.