Learn More
MP BIOMEDICALS Nuc-Off Nucleases and DNA Removal Spray
Non-toxic spray designed to remove nucleases, DNA, and RNA.
Brand: MP BIOMEDICALS 112462450
Description
MP Biomedicals Nuc-Off Nucleases and Nucleic Acid Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase, DNAase, RNA and DNA.
This non-toxic spray-type solution can effectively remove nucleases, RNA and DNA from the surface of:
- laboratory consumables
- instruments
- other utensils
creating a clean environment for RNA or DNA-related experiments.
It is strongly recommended to regularly clean your:
- pipettors
- bench surfaces
- instrument exterior
Application Notes:
Nuc-Off Nucleases and Nucleic Acid Removal Spray is intended for effective and safe elimination of Nucleases, DNA, and RNA that may cause false positive or false negative experimental results.
Usage Statement:
Research Use Only (RUO)
Specifications
Removal Agent | |
450 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.