Learn More
MP BIOMEDICALS Nuc-Off Nucleases and Nucleic Acid Removal Spray
Non-toxic spray designed to remove nucleases and DNA.
£178.00 - £676.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
30214853
|
MP BIOMEDICALS
112461450 |
450 mL |
£178.00
450mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30214848
|
MP BIOMEDICALS
112461451 |
1800 mL |
£676.00
1.80 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Biomedicals Nuc-Off Nucleases and DNA Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase, DNase and DNA contamination.
This non-toxic spray-type solution can effectively remove nucleases and DNA from the surface of:
- laboratory consumables
- instruments
- other utensils
creating a clean environment for RNA or DNA-related experiments.
It is strongly recommended to regularly clean your:
- pipettors
- bench surfaces
- instrument exterior
Application Notes:
Nuc-Off Nucleases and DNA Removal Spray is intended for effective and safe elimination of Nucleases and DNA that may cause false positive or false negative experimental results.
Usage Statement:
Research Use Only (RUO)
Specifications
Removal Agent |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.