Learn More
MP BIOMEDICALS Nuc-Off Nucleases Removal Spray
Non-toxic spray designed to remove nucleases.
Brand: MP BIOMEDICALS 112460451
Description
MP Biomedicals Nuc-Off Nucleases Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase and DNase.
This non-toxic and non-corrosive spray-type solution can effectively remove nucleases from the surface of:
- laboratory consumables
- instruments
- other utensils
- creating a clean environment for RNA or DNA-related experiments.\
It is strongly recommended to regularly clean your:
- pipettors
- bench surfaces
- instrument exterior
Application Notes:
Nuc-Off Nucleases Removal Spray is intended for effective and safe elimination of Nucleases that may cause false positive or false negative experimental results.
Usage Statement:
Research Use Only (RUO)
Specifications
Removal Agent | |
1800 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.