missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ NucGreen™ Dead 488 ReadyProbes™ Reagent (SYTOX™ Green)

NucGreen™ Dead 488 ReadyProbes™ Reagent

Brand:  Molecular Probes™ R37109

192.28 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15164175

  • £253.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

NucGreen Dead 488 ReadyProbes Reagent is a cell-impermeant stain that emits exceptionally bright, green fluorescence when bound to DNA. Cells that have lost plasma membrane integrity are stained within minutes, making this an extremely useful stain to estimate live/dead cell ratios and to measure cytotoxicity in kinetic live-cell assays. It is also suitable for staining nuclei in fixed cell preparations and tissue sections.

• Exceptionally bright green fluorescence upon binding to DNA
• Rapid staining of damaged, dead, or fixed cells without wash steps
• Ready-to-use liquid formulation in convenient dropper bottle—no need to dilute, weigh, or pipette
• Stability at room temperature—keep handy at your scope or cell

Cell imaging applications

The membrane-impermeable NucGreen Dead 488 reagent is a high-affinity DNA stain that easily stains cells with compromised cell membranes, yet does not enter living cells. It is therefore ideal for discrimination of live and dead cells. The fluorescence increases more than 500-fold on binding to dsDNA in cells with compromised cell membranes (dying or dead cells), in fixed cells, and tissue slices. The spectral properties, with excitation/emission at 504/523 nm when bound to DNA, are ideal for standard FITC filters.

Suggestions for use

• NucGreen Dead 488 reagent may be added directly to cells in full media or buffer solutions
• It easily stains nuclei of fixed cells and tissue slices
• In most cases, 2 drops/ml and an incubation time of 15 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained. In most cases, staining intensity increases with time if cells are not washed prior to imaging.
• NucGreen Dead 488 dye is excited with a maximum at 504 nm when bound to DNA, with an emission maximum at 523 nm. It is detected through standard GFP and FITC filters.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell-impermeant
6 x 2.5 mL dropper bottles

Store at ≤ 25°C
Viability Assay
Nucleic Acid Stain
523 nm
Liquid
6 Bottles
Green
Fluorescence
Confocal Microscope, Floid Cell Imaging System, Fluorescence Microscope, Flow Cytometer
Other Label(s) or Dye(s)
504 nm
NucGreen, ReadyProbes
Nucleus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.