Learn More
Macherey-Nagel™ NucleoBond™ High Molecular Weight DNA Kit
For large amounts of ultrapure, high molecular weight DNA fragments from cells and tissue
Brand: Macherey-Nagel™ 740160.20
178.88 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
Additional Details : Weight : 1.25000kg
Description
- High quality, high molecular weight (HMW) DNA up to 200 kbp
- Minimized DNA shearing due to established anion exchange technology
- Validated with diverse samples and sequencing platforms
Specifications
Midi Gravity-flow Column | |
Cell Cultures, Plant Leaves, Solid Tissue, Yeast, Bacteria, Liquid Sample (e.g., blood, body fluids or enzymatic reactions) | |
20 | |
120 min / 12 preps |
Anion Exchange Chromatography Technology | |
High molecular weight DNA Purification | |
Fragment size up to 200 kbp, > 90% depletion of fragments < 10kbp | |
Depending on the sample amount and type |
Safety and Handling
- NucleoBond HMW DNA (20)
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Research Use Only