missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ PC EF Kit Buffer Solutions

Additional supply of buffer for use with NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) for the purification of endotoxin-free plasmid DNA. Buffer solutions include: S1/S2/S3-EF, N2/N3/N4/N5-EF, TE-EF, and H2O-EF.

Brand:  Macherey-Nagel™ 740794.1


 View more versions of this product

Product Code. 15809117

  • £91.55 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NucleBond™ PC EF: The classic anion exchanger for purification of endotoxin-free plasmid DNA

  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Plasmid DNA with less than 0.1 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Fast and easy clarification of lysates by Folded Filters – no centrifugation required, no shearing forces
  • NucleoBond™ Finalizers available to avoid time consuming centrifugation for plasmid precipitation
  • Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Specifications

Specifications

Colorless
NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100)
6.0 to 6.5
64-17-5d
Room Temperature (18°C to 25°C)
Purification of endotoxin-free plasmid DNA
Ethanol, diluted < 20%; Potassium chloride; Water
Ethanol, diluted < 20%; Potassium chloride; Water
1000 mL
Alcoholic
Liquid
Wash Buffer N3-EF
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer N3-EF (1000 mL)

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, triton-x 114

SDS
Documents

Documents

Certificates
Special Offers

Special Offers