missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ PC EF Kit Buffer Solutions
Additional supply of buffer for use with NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) for the purification of endotoxin-free plasmid DNA. Buffer solutions include: S1/S2/S3-EF, N2/N3/N4/N5-EF, TE-EF, and H2O-EF.
Brand: Macherey-Nagel™ 740798.1
This item is not returnable.
View return policy
Description
NucleBond™ PC EF: The classic anion exchanger for purification of endotoxin-free plasmid DNA
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Plasmid DNA with less than 0.1 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Fast and easy clarification of lysates by Folded Filters – no centrifugation required, no shearing forces
- NucleoBond™ Finalizers available to avoid time consuming centrifugation for plasmid precipitation
- Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Specifications
Colorless | |
6.0 to 8.0 | |
64-17-5d | |
Room Temperature (18°C to 25°C) | |
Purification of endotoxin-free plasmid DNA |
NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) | |
1000 mL | |
Odorless | |
Liquid | |
Water |