missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Maxi kit for transfection-grade plasmid DNA
Click to view available options
No. of Reactions:
10
50
100
Unit Size:
0.1kg
100 preps
50 preps
Description
Features
- Optimized column design—Silica resin packed between filters, with prep in just 75 minutes
- Column filter included—High filter flow rates, parallel lysate clearing, and loading onto the column
- Improved silica material—Typical yields of 1,000μg and can be used over a wide pH range
- Proven anion-exchange technology—Transfection-grade plasmid DNA
- Kits contain: NucleoBond™ Xtra Maxi columns and filter, RNase A, buffers (RES, LYS, NEU, EQU, WASH, ELU) and plastic washers
Applications
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Specifications
| Buffer | Yes |
| Format | Maxi Gravity-flow Column |
| Isolation Technology | Anion Exchange Chromatography Technology |
| No. of Reactions | 50 |
| RNase | Yes |
| Sample Volume | <600 mL (high copy), <1200 mL (low copy) |
| Size | <300 kbp |
| Test Time | 75 min./prep |
| Yield | 1000μg |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction