missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Maxi Plus

Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Plus Kit represents the new generation of anion exchanger allowing high yield, ultra-fast. The Plus kit contains NucleoBond™ Finalizers to speed up DNA precipitation.

Brand:  Macherey-Nagel™ 740416.2

82.18 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.



Code : X3

Additional Details : Weight : 0.31000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 11915452

  • £86.50 / 2 reactions
Estimated Shipment: 02-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Features
  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design –  Maxi prep in 35 min
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 1000 μg (Maxi)
  • NucleoBond™ Xtra EF kits contain NucleoBond™ Finalizers to speed up DNA precipitation
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications

Specifications

Yes
Anion Exchange Chromatography Technology
2
< 600, < 1200μL
1000μg
Maxi Gravity-flow Column
Nucleo Bond Xtra Maxi Plus
Yes
<300kbp
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <

SDS
Documents

Documents

Certificates
Special Offers

Special Offers