missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Midi
Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Midi Kit represents the new generation of anion exchanger allowing high yield, ultra-fast.
Brand: Macherey-Nagel™ 740410.10
This item is not returnable.
View return policy
Description
Features
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Midi prep in 30 min.
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 400μg (Midi)
Applications
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Midi Gravity-flow Column | |
10 | |
400µg |
Anion Exchange Chromatography Technology | |
Midi prep |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <