missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Midi kit for transfection-grade plasmid DNA

Product Code. 12798402
Click to view available options
No. of Reactions:
10
50
100
Unit Size:
10 reactions
100 preps
50 preps
This item is not returnable. View return policy

Product Code. 12798402

Brand: Macherey-Nagel™ 740410.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Midi Kit represents the new generation of anion exchanger allowing high yield, ultra-fast.

Features
  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design – Midi prep in 30 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 400μg (Midi)
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)

Specifications

Buffer Yes
Format Midi Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
No. of Reactions 50
RNase Yes
Sample Volume <200 mL (high copy), <400 mL (low copy)
Size <300 kbp
Yield 400μg
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.