missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Nucleodur™ C18 Analytical EC Standard Column
Use EC standard columns for analytical HPLC (High Performance Liquid Chromatography). Nucleodur™ C18 Analytical EC Standard Columns come in a wide variety of lengths and internal diameters.
£448.00 - £785.00
Specifications
Column Chemistry | Silica |
---|---|
Column Format | Analytical Column |
Crosslinking | Octadecyl |
For Use With | HPLC |
Mobile Phase Compatibility | Acetonitrile/water |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Carbon Load | Particle Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Carbon Load | Particle Size | Price | Quantity & Availability | ||
10376732
|
Macherey-Nagel™
760201.20 |
2 mm | 125 mm | 14% | 5 μm |
£520.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12468367
|
Macherey-Nagel™
760103.20 |
2 mm | 150 mm | 18% | 5 μm |
£520.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12478357
|
Macherey-Nagel™
760100.20 |
2 mm | 125 mm | 18% | 5 μm |
£520.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12418367
|
Macherey-Nagel™
760101.20 |
2 mm | 250 mm | 18% | 5 μm |
£605.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12478387
|
Macherey-Nagel™
760203.20 |
2 mm | 150 mm | 14% | 5 μm |
£520.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12438367
|
Macherey-Nagel™
760102.20 |
2 mm | 50 mm | 18% | 5 μm |
£448.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12458387
|
Macherey-Nagel™
760202.20 |
2 mm | 250 mm | 14% | 5 μm |
£605.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12488357
|
Macherey-Nagel™
760100.30 |
3 mm | 125 mm | 18% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
10091852
|
Macherey-Nagel™
760201.30 |
3 mm | 125 mm | 14% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12478367
|
Macherey-Nagel™
760103.30 |
3 mm | 150 mm | 18% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12408387
|
Macherey-Nagel™
760200.20 |
2 mm | 50 mm | 14% | 5 μm |
£448.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
10530910
|
Macherey-Nagel™
760101.30 |
3 mm | 250 mm | 18% | 5 μm |
£600.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12488387
|
Macherey-Nagel™
760203.30 |
3 mm | 150 mm | 14% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12468387
|
Macherey-Nagel™
760202.30 |
3 mm | 250 mm | 14% | 5 μm |
£600.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12448367
|
Macherey-Nagel™
760102.30 |
3 mm | 50 mm | 18% | 5 μm |
£448.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12728722
|
Macherey-Nagel™
760104.46 |
4.6 mm | 100 mm | 18% | 5 μm |
£580.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12418387
|
Macherey-Nagel™
760200.30 |
3 mm | 50 mm | 14% | 5 μm |
£448.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12737810
|
Macherey-Nagel™
760204.46 |
4.6 mm | 100 mm | 14% | 5 μm |
£580.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12408367
|
Macherey-Nagel™
760100.46 |
4.6 mm | 125 mm | 18% | 5 μm |
£580.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12448387
|
Macherey-Nagel™
760201.46 |
4.6 mm | 125 mm | 14% | 5 μm |
£580.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
10860121
|
Macherey-Nagel™
760103.46 |
4.6 mm | 150 mm | 18% | 5 μm |
£580.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12408397
|
Macherey-Nagel™
760203.46 |
4.6 mm | 150 mm | 14% | 5 μm |
£580.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12428367
|
Macherey-Nagel™
760101.46 |
4.6 mm | 250 mm | 18% | 5 μm |
£635.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
10586093
|
Macherey-Nagel™
760202.46 |
4.6 mm | 250 mm | 14% | 5 μm |
£635.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
10599552
|
Macherey-Nagel™
760102.46 |
4.6 mm | 50 mm | 18% | 5 μm |
£491.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12438387
|
Macherey-Nagel™
760200.46 |
4.6 mm | 50 mm | 14% | 5 μm |
£491.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12428407
|
Macherey-Nagel™
760262.46 |
4.6 mm | 250 mm | 14% | 3 μm |
£785.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12738722
|
Macherey-Nagel™
760106.46 |
4.6 mm | 75 mm | 18% | 5 μm |
£491.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12747810
|
Macherey-Nagel™
760205.46 |
4.6 mm | 75 mm | 14% | 5 μm |
£491.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12498357
|
Macherey-Nagel™
760100.40 |
4 mm | 125 mm | 18% | 5 μm |
£525.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
10001862
|
Macherey-Nagel™
760201.40 |
4 mm | 125 mm | 14% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12488367
|
Macherey-Nagel™
760103.40 |
4 mm | 150 mm | 18% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12498387
|
Macherey-Nagel™
760203.40 |
4 mm | 150 mm | 14% | 5 μm |
£525.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
10787774
|
Macherey-Nagel™
760101.40 |
4 mm | 250 mm | 18% | 5 μm |
£600.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
12458367
|
Macherey-Nagel™
760102.40 |
4 mm | 50 mm | 18% | 5 μm |
£448.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
10163462
|
Macherey-Nagel™
760202.40 |
4 mm | 250 mm | 14% | 5 μm |
£600.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12428387
|
Macherey-Nagel™
760200.40 |
4 mm | 50 mm | 14% | 5 μm |
£448.00
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
Description
Key features:- Stable in 100 % aqueous mobile phase systems
- Interesting polar selectivity features
- Excellent base deactivation; suitable for LC/MS due to low bleeding characteristics
- Special phase with polar endcapping
- Pore size 110 Å
- Particle sizes 1.8 μm, 3 μm and 5 μm
- 7 and 10 μm particles for preparative purposes on request
- Carbon content 14 %
- pH stability 1–9
Recommended application:
- Analgesics, penicillin antibiotics, nucleic acid bases, water-soluble vitamins, complexing agents, organic acids
Specifications
Silica | |
Octadecyl | |
Acetonitrile/water | |
1 Ea. | |
L1 | |
Reversed Phase | |
Reversed Phase |
Analytical Column | |
HPLC | |
110 Å | |
Acetonitrile/water | |
Silica | |
Yes | |
Analytical EC Column |