missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nucleodur™ C8 ec VariPerp HPLC Column

Column Format: Preparative Column; Carbon Load: 10.50%; Pore Size: 110 Å; Column Phase: Reversed Phase

Brand:  Macherey-Nagel™ 762062.100

Code : 16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12419906

  • £880.00 / Each
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Preparative Column
Basic, Neutral or Acidic Drugs, Derivatized Amino Acids, Pesticides, Fat-Soluble Vitamins, Aldehydes and Ketones, Phenolic Compounds
5800 psi (400 bar)
1 to 9
340 m²/g
L7
Silica
Reversed Phase
Reversed Phase
10 mm
250 mm
5 μm
110 Å
60°C
C8
10.5%
Yes
VariPerp HPLC Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers