Learn More
Macherey-Nagel™ Nucleodur™ CN-RP HPLC CC Guard Columns
Brand:  Macherey-Nagel™ 761420.40
 View more versions of this product
Product Code. 12438577
  • £139.00 / Each
Estimated Shipment: 05-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
7%
8 mm
CC Guard Column
NUCLEODUR™ CN-RP
Acetonitrile/Water
Cyano
Tricyclic Antidepressants, Steroids, Organic Acids
Spherical
L10
5 μm
100Å
4 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
Yes