Learn More
Lonza™ Nucleofector™ Transfection 2b Device
Used for bacteria transformation by using alternative cuvettes. Lonza Nucleofector™ Transfection 2b Device has over 150 ready-to-use Amaxa™ Optimized Protocols containing cell type specific guidance.
Brand: Lonza™ AAB-1001
Additional Details : Weight : 1.00000kg
Description
- Highly efficient transfection of primary cells and cell lines
- Reliable results due to high viability and preservation of cell functionality
Applications:
- Low-throughput transfection in single cuvette format
- Transfection of plasmid DNA, siRNA, shRNA, miRNA, RNA and morpholinos
- Transfection of peptides, proteins or small molecules
- Approaching 4,000 peer-reviewed publications
- Suited for bacteria transformation
Specifications
Transfection 2b Device | |
100 to 110V |