missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Nucleon™ BACC3 Genomic DNA Extraction Kit

Nucleon BACC DNA Extraction kits use a Nucleon resin to bind proteins ensuring high yields of high-quality, high molecular weight genomic DNA in only one hour

Brand:  Cytiva RPN8512

384.75 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.



Product Code. 10004244

  • £409.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Reagents A and B, 5.0M Sodium Perchlorate, and Nucleon resins.
This item is not returnable. View return policy

Description

Description

  • Phenol-free protocol requiring only 30 minutes to complete
  • Low-shearing protocol gives maximum recoveries of high-molecular weight DNA
  • For 50 preps of 10mL of whole blood (sufficient quantities of all reagents provided)
  • Available in different formats/reaction scales (see 45-000-955 and 45-000-956)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

illustra Nucleon BACC system rapidly extracts high molecular weight DNA from whole blood and cultured cells.
Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood.
illustra Nucleon BACC3
370-440 μg DNA/mg tissue
Blood, Cell Cultures
Reagents A and B, 5M Sodium Perchlorate and Nucleon™ Resins
50 Kits
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising liquid Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Supplemental information
  • MIXTURE LIST-Contains : potassium nitrate, Poly(oxy-1, 2-ethanediyl),3-tetramethylbutyl)phenyl]-

SDS
Documents

Documents

Certificates
Special Offers

Special Offers