missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Bioanalysis™ Nucleospin™ Dx Virus

CE-IVD marked mini spin kit for the isolation of viral RNA and DNA

Brand:  Macherey Nagel Bioanalysis™ 740895.50


Product Code. 11992382

  • £199.00 / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

contains Dx virus columns with collection tubes, lysis tubes (1.5mL), collection tubes (2mL), elution tubes (1.5mL), buffers, Proteinase K, Carrier RNA 50 preps Macherey-Nagel
Specifications

Specifications

18°C to 25°C
Plasma, Serum
50 Preps
100 bp−approx. 50 kbp
Mini Prep
Isolation of Viral Nucleic Acids
150 L
Viral RNA and DNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.