missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Food, Mini kit for DNA from food
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
Description
Features:
- For rapid small-scale isolation of genomic DNA from food and feed
- Complete removal of PCR inhibitors – get high quality DNA
- Even low amounts of partially degraded DNA can be purified from complex matrices
- Fast and easy procedure DNA from various sample materials – highest flexibility
Applications:
- DNA from complex matrices, e.g., processed food, soy (milk and flour), chocolate, cereals, meat, animal feed
- Typical downstream applications: real-time PCR, Southern blotting, enzymatic reactions
Specifications
Specifications
| Detection Method | Silica-membrane Technology |
| Format | Mini Spin Column |
| For Use With (Equipment) | PCR, Southern Blotting, Enzymatic Reactions |
| Includes | 2mL Collection Tubes, Buffers, Proteinase K |
| No. of Reactions | 10 Preps |
| Product Type | DNA Isolation Columns |
| Quantity | 1 Each |
| Sample Volume | <200 mg |
| Test Time | 30 min. |
| Yield | 0.1 to 10 μg (200 mg food) |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : guanidine hydrochloride/dodecyl sulfate,sodium salt/ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction