missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Food, Mini kit for DNA from food

Product Code. 11972402
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 11972402

Brand: Macherey-Nagel™ 740945.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate genomic DNA from food and feed. NucleoSpin™ Food Columns are perfect for small-scale DNA isolation for a variety of downstream applications.

Features:

  • For rapid small-scale isolation of genomic DNA from food and feed
  • Complete removal of PCR inhibitors – get high quality DNA
  • Even low amounts of partially degraded DNA can be purified from complex matrices
  • Fast and easy procedure DNA from various sample materials – highest flexibility

Applications:

  • DNA from complex matrices, e.g., processed food, soy (milk and flour), chocolate, cereals, meat, animal feed
  • Typical downstream applications: real-time PCR, Southern blotting, enzymatic reactions

Specifications

Detection Method Silica-membrane Technology
Format Mini Spin Column
For Use With (Equipment) PCR, Southern Blotting, Enzymatic Reactions
Includes 2mL Collection Tubes, Buffers, Proteinase K
No. of Reactions 50 Preps
Product Type DNA Isolation Columns
Quantity 1 Each
Sample Volume <200 mg
Test Time 30 min.
Yield 0.1 to 10 μg (200 mg food)
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride/dodecyl sulfate,sodium salt/ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.