Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Columns
Replacement mini spin columns for use with NucleoSpin™ Gel and PCR Clean-up kits to allow DNA fragments to be purified from enzymatic reactions, such as PCR, as well as from agarose gels.
Brand: Macherey-Nagel™ 740609.50S
52.49 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
NucleoSpin™ Gel and PCR Clean-up Kit — Two-in-one kit with optimized recovery and elution volume
- Two applications in one kit – PCR clean-up and gel extraction
- Uses one buffer with optimal performance for both applications
- High recoveries for fragments down to 50 bp
- Minimized elution volume of 15 μL – highly concentrated DNA
- Binding buffer with pH indicator
- Separate buffers for single stranded DNA or SDS containing samples available
- Suitable for all gel buffer systems (e.g., TAE, TBE)
Specifications
Mini Spin Column | |
Silica Membrane | |
50 Pk. |
NucleoSpin Gel and PCR Clean-up Kit | |
Spin Column |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Supplemental information
- NucleoSpin Gel and PCR Clean-up
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate
Product Content Correction
Product Title
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Columns > 50 columns
Spot an opportunity for improvement?Share a Content Correction