missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Columns
Clean up PCR Columns for use with NucleoSpin kits.
Brand: Macherey-Nagel™ 740609.250S
231.22 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
- Columns only (no buffers) for PCR purification or gel extraction applications.
- In accordance with Macherey-Nagel´s Quality Management System, each lot of product is tested against predetermined specifications to ensure consistent product quality.
Specifications
Mini Spin Column | |
Typical recovery: 70–95% | |
For use with NucleoSpin kits. | |
250 Columns | |
<400μL (PCR reaction mixture) <400mg (TAE / TBE agarose gel) |
|
10 min/6 preps |
Silica Membrane Technology | |
Elution volume: 15–30μL | |
NucleoSpin Gel and PCR Clean-up Columns (250) | |
A260/A280: 1.8–1.9 | |
50 bp to ∼20 kbp | |
Up to ∼15μg DNA |
Safety and Handling
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate