missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up XS Columns
Columns for concentrated clean up with small elution volumes
Brand: Macherey-Nagel™ 740611.50S
Additional Details : Weight : 0.23520kg
Description
- Rapid concentration and clean up of PCR products
- 2 in 1 column for reaction mixtures and gel extraction
- XS column design enables elution of highly concentrated DNA
Specifications
5 µg | |
Non hazardous, 18°C to 25°C | |
Silica Membrane Technology | |
DNA | |
Agarose gel | |
No | |
Gel Extraction and PCR Clean-up | |
DNA | |
A260/A280: 1.8-1.9 | |
Room Temperature | |
DNA Fragments | |
Recovery 70-95% |
XS Spin Column | |
Spin Column - XS Design | |
6 to 12 µL | |
Centrifugation, Vacuum | |
PCR Clean-up and Gel Extraction, Sample Concentration | |
NucleoSpin Gel and PCR Clean-up XS Columns Only | |
50 | |
XS Column | |
<200 µL PCR reaction mixture, <200 mg agarose gel | |
50 bp-20 kbp | |
15 min./6 Preps |
For research use only