missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ miRNA Plasma

Complete kit for the isolation of small RNA and DNA from plasma and serum samples.

Brand:  Macherey-Nagel™ 740981.50

263.96 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.



Product Code. 12375403

  • £345.00 / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Efficient and selective isolation of small RNA from plasma or serum samples
  • Superior recovery of small RNA ≥ 18 nt
  • Optional on-column DNA digest for increased sensitivity in downstream applications
  • Simple and fast procedure – without phenol / chloroform
  • Ideal for use in cancer research (e.g., reliable monitoring of clinical parameters
  • Typical downstream applications: real-time qRT-PCR, chip hybridisations
Specifications

Specifications

Green band
Mini Spin Column
20 - 50 μL
Bindding capacity: 200 μg
Centrifuge
NucleoSpin miRNA Columns, Collection Tubes (2 mL), buffers, RNase-free rDNase
300 μL
rDNase at 4 °C; Other components at room temperature (18 - 25 °C)
Silica Membrane
Plasma, Serum
Small RNA and DNA
Isolation of nucleic acids
50 Preps
40 min/10 preps (without rDNase digestion); 70 min/10 preps (with rDNase digestion)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.