missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plant II

Isolate and purify genomic DNA from plant samples. NucleoSpin™ Plant II kits enable enhanced yield and quality DNA.

Brand:  Macherey-Nagel™ 740770.10

42.39 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 11992492

  • £62.10 / 10 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • The second generation for rapid isolation of genomic DNA from plant samples
  • with higher yield and quality
  •  Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
  • Lysis Buffer PL1, based on CTAB lysis method
  • Lysis Buffer PL2, based on SDS lysis method
  • NucleoSpin™ Filters included for clarification of lysate
  • Improved buffer compositions – higher yield and purity
  • Optimized silica membrane – improved DNA binding
  • RNase A included
Applications
  • DNA from plant cells and tissue
  • DNA from fungi
  • Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications

Specifications

10 Preps
Plant II
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride, ethanol/dodecylsulfate, sodium salt/guanidine hydrochloride, 2-propanol

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers