missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid Columns

Replacement columns for use with NucleoSpin™ Plasmid kits for rapid, small-scale preparation of high-purity, sequencing-grade plasmid DNA.

£60.75 - £293.00

Specifications

Color Clear
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 50 μg
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15870928
View Documents
Macherey-Nagel™
740588.50S
50 Each
£60.75
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
15860928
View Documents
Macherey-Nagel™
740588.250S
250 Each
£293.00
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NucleoSpin™ Plasmid

  • For rapid small scale preparation high-purity plasmid DNA
  • Fast procedure – 6 Mini preps in 25 min
  • High capacity – up to 50 μg of plasmid DNA
  • Chaotropic salt wash buffer – extra pure DNA
  • Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
    • High-copy plasmids from E. coli
    • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
    • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
    • Supplementary protocol for M13 DNA availabl
Specifications

Specifications

Clear
Silica Membrane Technology
Binding capacity: 50 μg
Sequencing-grade plasmid DNA
Preparation of high-purity plasmid DNA
NucleoSpin Plasmid lidded Column
Mini Spin Column
E. coli Culture
Elution volume: 50 μL
NucleoSpin™ Plasmid Kit
Columns only
2 - 10 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NucleoSpin Plasmid

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt/guanidine hydrochloride/2-pr

SDS
Documents

Documents

Product Certifications