Learn More
Macherey-Nagel™ NucleoSpin™ Plasmid
Prepare small scale high-purity plasmid DNA rapidly. NucleoSpin™ Plasmid comes with or without a cap lid.
Brand: Macherey-Nagel™ 740588.50
81.77 GBP valid until 2024-12-31
Use promo code "21949" to get your promotional price.
Code : 86
Additional Details : Weight : 0.41400kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
Features:
- For rapid small scale preparation high-purity plasmid DNA.
- Fast procedure – 6 Minipreps in 25 min.
- High capacity – up to 50μg of plasmid DNA.
- Chaotropic salt wash buffer – extra pure DNA.
Applications:
- High-copy plasmids from E. coli.
- Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required).
- Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures.
- Supplementary protocol for M13 DNA available on request.
- Typical downstream applications: cloning, sequencing, PCR, transformation, restriction analysis.
Specifications
Column | |
DNA and RNA Sequencing | |
Lid | |
Plasmid Columns | |
2 to 10 mL | |
25 min. |
Silica Membrane Technology | |
High-copy plasmids from E. coli | |
50 Preps | |
2 mL | |
<50 kbp |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt/guanidine hydrochloride/2-pr