missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid DNA

Product Code. 11942362
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Includes:
Lid
No Lid
Unit Size:
10 reactions
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 11942362

Brand: Macherey-Nagel™ 740499.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepare small scale high-purity plasmid DNA rapidly. NucleoSpin™ Plasmid Columns come with or without a cap lid.

Features:

  • For rapid small scale preparation high-purity plasmid DNA
  • Fast procedure – 6 Minipreps in 25 min.
  • High capacity – up to 50μg of plasmid DNA
  • Chaotropic salt wash buffer – extra pure DNA

Applications:

  • High-copy plasmids from E. coli
  • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
  • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
  • Supplementary protocol for M13 DNA available on request
  • Typical downstream applications: cloning, sequencing, PCR, transformation, restriction analysis

Specifications

Format Column
Isolation Technology Silica Membrane Technology
Sequencing Type DNA and RNA Sequencing
For Use With (Application) High-copy plasmids from E. coli
Includes No Lid
No. of Reactions 50 Preps
Product Type Plasmid Columns
Quantity 2 mL
Sample Volume 2 to 10 mL
Size <50 kbp
Test Time 25 min.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %); guanidine hydrochloride; 2-propanol;Rnase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.