missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Plasmid EasyPure, Mini kit for easy plasmid DNA purification
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 preps
Pack of 250
Pack of 50
Specifications
Specifications
| No. of Reactions | 50 Preps |
| Product Type | Plasmid EasyPure |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- MIXTURE LIST-Contains : dodecyl sulfate, sodium salt, guanidinehydrochloride, sodium hydroxide solution, RNase liquid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction