missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid EasyPure, Mini kit for easy plasmid DNA purification

Product Code. p-8013753
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 preps
Pack of 250
Pack of 50
This item is not returnable. View return policy

Product Code. 15266437

Brand: Macherey-Nagel™ 740727.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultrafast and ease of use Mini preparation of plasmid DNA.

Specifications

No. of Reactions 10 Preps
Product Type Plasmid EasyPure
compliance-icons
Product Identifier
  • NucleoSpin Plasmid EasyPure
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), guanidine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.