missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin Plasmid QuickPure™ Kit

Features a specially treated silica membrane that speeds up the procedure by combining washing and drying into one step. Macherey-Nagel™ NucleoSpin Plasmid QuickPure™ provides ultra-fast small-scale plasmid DNA purification which offers a saving of time and costs.

£98.40 - £468.00

Specifications

Content And Storage Room temperature (18–25°C)
Format Mini Spin Column
For Use With (Equipment) Standard benchtop microcentrifuge
For Use With (Application) Plasmid DNA purification/isolation
Includes Resuspension Buffer A1, Lysis Buffer A2 (without LyseControl), Neutralization Buffer A3, Wash Buffer AQ (Concentrate), Elution Buffer AE, RNase A (lyophilized), NucleoSpin™ Plasmid QuickPure Columns (dark yellow rings), Collection Tubes (2mL), User manual
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
11902422
View accessory products View Documents
Macherey-Nagel™
740615.250
250 Preps This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11902362
View accessory products View Documents
Macherey-Nagel™
740615.50
50 Preps This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

With the NucleoSpin™ Plasmid QuickPure method, the bacteria culture is lysed according to alkaline lysis. The lysate is cleared by centrifugation and the supernatant is loaded onto the NucleoSpin™ Plasmid QuickPure column. Contaminations like salts, metabolites, nucleases and soluble macromolecular cellular components are removed by only one single washing step with buffer AQ. Pure plasmid DNA is finally eluted under low ionic strength conditions with slightly alkaline buffer AE (5mM Tris-Cl, pH 8.5). The prepared plasmid DNA is suitable for all common downstream applications.

Kit components:
Resuspension Buffer, Lysis Buffer, Neutralization Buffer, Wash Buffer, Elution Buffer, RNase A, NucleoSpin™ Plasmid QuickPure Columns, Collection Tubes, User manual

Features:

  • Yield: up to 15μg
  • Ultra-fast procedure: 18 minipreps in 11min
  • Elution volume: 50μL
  • Sample material: 1mL to 3mL E. coli culture
  • Very good price-performance ratio
  • Silica-membrane technology
  • Binding capacity: 15μg
  • No use of organic solvents
  • Ultra-fast small scale preparation of plasmid DNA
  • Highly pure nucleic acids suitable for all common downstream applications
  • Vectors <15kb
  • Format: mini-spin columns
For further information on this product please contact Customer Services.
Specifications

Specifications

Room temperature (18–25°C)
Standard benchtop microcentrifuge
Resuspension Buffer A1, Lysis Buffer A2 (without LyseControl), Neutralization Buffer A3, Wash Buffer AQ (Concentrate), Elution Buffer AE, RNase A (lyophilized), NucleoSpin™ Plasmid QuickPure Columns (dark yellow rings), Collection Tubes (2mL), User manual
Lyophilized
<15 kbp
<15μg (1–3mL culture)
Mini Spin Column
Plasmid DNA purification/isolation
NucleoSpin Plasmid QuickPure
Culture volume: 1–3mL high copy
Prep time: 11min./18 preps
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1Year

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt

SDS
Documents

Documents

Product Certifications