missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid Transfection-grade, Mini kit for ultrapure plasmid DNA

Product Code. 15593626
Click to view available options
No. of Samples:
50
250
Unit Size:
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 15593626

Brand: Macherey-Nagel™ 740490.250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A fast way to purify transfection-grade plasmid DNA using mini-format technology to reduce endotoxins from bacterial lysates. Complete kit includes mini spin columns for an ultra-fast workflow.

Endotoxins are co-purified during typical plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. NucleoSpin™ Plasmid Transfection-grade kits are designed to reduce endotoxins from bacterial lysates.

  • Purifies high-copy plasmids for transfection
  • Novel technology to diminish endotoxin content (< 50 EU/μg DNA) for successful transfection
  • Rapid purification from E. coli via manual, vacuum or centrifugation processing
  • Ultra-fast procedure using mini spin columns simplifies the work-flow
  • Downstream applications: transfection of common cell lines (lipofection, electroporation)
  • Also suitable for standard molecular biology application, such as cloning and sequencing

Specifications

Bacterial Endotoxins <50 EU/ng DNA
Content And Storage Room temperature (18 - 25 °C)
Format Mini Spin Column
Isolation Technology Silica Membrane and Endotoxin-reduction Technology
Sample Type Bacterial Culture (E. coli, High-copy Plasmids)
Efficiency Binding capacity: 35 μg
Elution Volume Elution Volume: 30 - 50 μL
Final Product Type Transfection-grade plasmid DNA
For Use With (Equipment) Centrifuge, Vacuum manifold (e.g. NucleoVac 96)
For Use With (Application) Rapid purification of plasmids for transfection
Includes NucleoSpin Plasmid Transfection-grade Columns, Collection Tubes, Buffers, RNase A
No. of Samples 250
Purity A260/A280: 1.80 - 1.85
Sample Volume Up to 5 mL
Size < 15 kbp
Test Time 14 min/6 preps
Yield 15 - 30 μg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt/guanidine hydrochloride/trie
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.