missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Blood

Isolate RNA with a simple and convenient spin column format. Macherey Nagel™ NucleoSpin™ RNA Blood is optimized for isolation of RNA from fresh or frozen whole blood (human or animal).

Brand:  Macherey-Nagel™ 740200.50

Code : NEW

Additional Details : Weight : 0.12000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 12733391

  • £277.00 / 120mL
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

 Features
  • Direct total blood lysis – very simple and convenient procedure (patent pending)
  • No selective erythrocyte lysis at 4 °C – complete processing at room temperature
  • Superior RNA yield and quality from up to 400 μL whole blood
  • Efficient on-column DNA removal for an increased sensitivity in downstream applications Compatible with common blood collection tubes and anticoagulants 
  •  (e.g., EDTA, citrate, and heparin)
Applications
  • Isolation of RNA from fresh or frozen whole blood from common blood collection tubes 
  •   (EDTA, citrate, heparin)
  • Typical downstream applications: qRT-PCR, next generation sequencing, blotting, array technology
Specifications

Specifications

Mini Spin Column
NucleoSpin™ RNA Blood Columns with Collection Tubes, Collection Tubes (2 mL), buffers, RNase-free rDNase, Liquid Proteinase K
NucleoSpin RNA Blood
>200 nt
1 to 8μg
Isolation of RNA from fresh or frozen whole blood from common blood collection tubes
50
Yes
55 min./6 Reaction
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate,ethanol (diluted< 20 %) /guanidinehydrochloride,ethanol/

SDS
Documents

Documents

Certificates
Special Offers

Special Offers