Learn More
Macherey-Nagel™ NucleoSpin™ RNA Clean-up XS Column
Brand: Macherey-Nagel™ 740903.50
231.87 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
Code : LE
Additional Details : Weight : 0.50000kg
Specifications
One-phase extraction | |
NucleoSpin™ RNA Clean-up XS Column | |
Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers | |
NucleoSpin™ RNA Clean-up XS Column | |
Reagent |
One-phase Extraction | |
For RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms | |
50 | |
1 Each |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate