missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Plus

State-of-the-art, dual-column kit includes gDNA removal columns and RNA binding columns for efficient and quick RNA isolation from cells and tissues.

Brand:  Macherey-Nagel™ 740984.250

1327.37 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



Product Code. 15300205

  • £2175.00 / 250 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NucleoSpin™ RNA Plus

  • Efficient and quick RNA isolation from cells and tissues
  • Two-column system includes gDNA removal columns and RNA binding columns
  • NucleoSpin™ gDNA Removal Columns – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal on one column in one step
  • New efficient lysis buffer – no b-mercaptoethanol or TCEP necessary
  • Variety of sample materials – cultured cells, animal tissue, plant tissue, microorganisms
  • Downstream applications include real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications

Specifications

Yellow/Blue ring
Mini Spin Column
30 - 120 μL
Binding capacity: 200 μg
Centrifuge
NucleoSpin RNA Plus Columns, NucleoSpin gDNA Removal Columns, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers
RNA Isolation Kit
< 30 mg tissue; < 30 mg tissue
HeLa cells (5 x 106): 40-60 μg
Mouse liver (20 mg): 80-100 μg
Mouse kidney (20 mg): 40-70 μg
Mouse spleen (5 mg): 30-60 μg
Room temperature (18-25 °C)
Silica Membrane
Cultured Bacterial or Yeast Cells, Human or Animal Tissue
RNA (with typical RIN > 9)
Isolation of RNA
250 Preps
A260/A280: 1.9 - 2.1
20 min/6 preps
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Supplemental information
  • MIXTURE LIST-Contains : ethyleneglycol diacetate, guanidinium thiocyanate, guanidinehydrochloride, ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers