Learn More
Macherey Nagel Bioanalysis™ NucleoSpin™ RNA Plus Isolation Column
For use in RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms, RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays. Macherey Nagel Bioanalysis NucleoSpin™ RNA Plus renders lysate clearin
Brand: Macherey Nagel Bioanalysis™ 740984.50
304.49 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.63000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
- rDNase digestion
- Typical RIN (RNA integrity number) > 9
- Elution volume: 30–120μL
- Preparation time: 20min/6 preps
- Binding capacity 200μg
Specifications
Lysis Buffer | |
Elution volume: 30–120 μL | |
RNA isolation from cells and tissues | |
50 | |
Typical yield: HeLa cells 5 x 106: 40-60μg; Mouse liver 20mg: 80-100μg; Mouse kidney 20mg: 40-70μg; Mouse spleen 5mg: 30-60μg |
Mini Spin Column | |
<1 x 107 Cultured Cells, <1 x 109 Bacterial Cells, <1 x 108 Yeast Cells, <30 mg Human/Animal Tissue | |
NucleoSpin gDNA removal Column | |
RNA isolation kit |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : ethyleneglycol diacetate / guanidinium thiocyanate /guanidine hydrochloride, ethanol
Kits to be used for research purposes only