Learn More
Macherey-Nagel™ NucleoSpin™ RNA Set for NucleoZOL
RNA purification from cells and tissue with NucleoZOL lysis and well-known NucleoSpin™ RNA Columns.
Brand: Macherey-Nagel™ 740406.50
Additional Details : CAS Number : 123-91-1 Weight : 0.38000kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
Combines NucleoZOL efficiency with proven NucleoSpin™ purity
- Simple bind-wash-elute procedure
- Perfect recovery – no loss of total RNA, including miRNA
- Superior RNA yields due to the efficient lysis using NucleoZOL reagent
- Save time and benefit from the easy and proven handling
- Highest quality – even from difficult sample materials
- NucleoZOL reagent (man. part no. 740404.200) sold separately
Specifications
Colorless | |
Mini Spin Column | |
NucleoZOL Lysis of Cell or Tissue Sample | |
Small RNA (10-200 nt) and large RNA (> 200 nt) | |
RNA purification | |
50 | |
500 μL NucleoZOL lysate | |
200 μg binding capacity |
Room temperature (18 - 25 °C) | |
Silica Membrane | |
85-95% recovery | |
NucleoZOL Reagent (740404.200) | |
Columns (50), Collection tubes (50 x 2 mL and 50 x 1.5 mL), Reagents (60 mL Binding Buffer MX, 12 mL Wash Buffer Concentrate RA3, 13 mL RNase-free Water) | |
Odorless | |
< 1 h |
Safety and Handling
- NucleoSpin RNA Set for NucleoZOL (50)
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : 1,4-dioxane