Learn More
Macherey-Nagel™ NucleoSpin™ RNA Virus Columns
Additional silica membrane mini spin columns for reliable purification of viral RNA from cell-free biological fluids (e.g., serum and plasma) using the NucleoSpin™ RNA Virus kit.
Brand: Macherey-Nagel™ 740956.250S
446.27 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
Additional Details : Weight : 0.65000kg
Description
NucleoSpin™ RNA Virus
- Reliable and reproducible isolation of viral RNA free of impurities
- Safe and well-established procedure
- Proven for a large variety of viruses including influenza viruses, HCV, HIV, Bird Flu Virus, Blue Tongue Virus
- Carrier RNA included for highest sensitivity in downstream applications
- Fast and convenient protocol – viral RNA and DNA* within only 30 min
- Support protocol for parallel isolation of viral RNA and DNA (with Proteinase K, not included in Kit)
- Viral RNA from 150 μL serum, plasma, cell-free biological fluids
- Typical downstream applications: RT-PCR, enzymatic reactions
Specifications
Blue band | |
Mini Spin Column | |
50 μL | |
Binding capacity: 40 μg | |
Isolation of viral RNA | |
250 Columns | |
30 min/4-6 preps |
Room temperature (18 - 25 °C) | |
Silica Membrane | |
Cell-free Biological Fluids, Plasma, Serum | |
Centrifuge | |
Columns only | |
< 150 μL |
Safety and Handling
- NucleoSpin RNA Virus
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, ethanol