missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA XS Column
Purify high quality RNA quickly and easily. NucleoSpin™ RNA XS Columns enable purification of highly concentrated RNA from the smallest samples
£135.00 - £1855.00
Specifications
Format | Mini Spin Column |
---|---|
Final Product Type | NucleoSpin™ RNA XS Column |
For Use With (Application) | For RNA isolation from cultured cells, tissue, cryosections and laser captured cells |
Includes | Collection tubes, NucleoSpin™ filters, collection tubes (2mL), Collection tubes (1.5mL), buffers, RNase-free rDNase, carrier RNA, reducing agent TCEP |
Product Type | NucleoSpin™ RNA XS Column |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12703580
|
Macherey-Nagel™
740902.10 |
10 |
£135.00
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12713580
|
Macherey-Nagel™
740902.250 |
250 |
£1855.00
250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12723580
|
Macherey-Nagel™
740902.50 |
50 |
£663.00
50 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features- Isolation of RNA from small sample quantities like biopsy material or single cells
- Excellent RNA recovery and integrity
- Concentrated RNA for sensitive downstream applications by elution in as little as 5 μL
- rDNase included for on-column DNA removal
- NucleoSpin™ Filters (shredders) – efficient homogenization and reduction of viscosity
- High quality RNA, ready to use for RT-PCR and other applications
- Reducing agent TCEP – no ß-mercaptoethanol necessary
Applications
- RNA isolation from cultured cells
- RNA isolation from tissue
- RNA isolation from cryosections
- RNA isolation from laser captured cells
- RNA isolation from small amounts of plant material
- RNA isolation from samples stored in RNAlater™
- Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology,
- RNase protection assays
Specifications
Mini Spin Column | |
For RNA isolation from cultured cells, tissue, cryosections and laser captured cells | |
NucleoSpin™ RNA XS Column | |
0.1 to 1.5ng (102 HeLa cells), 10 to 15ng (103 HeLa cells), 100 to 150ng (104 HeLa cells), 1000 to 1500ng (105 HeLa cells) |
NucleoSpin™ RNA XS Column | |
Collection tubes, NucleoSpin™ filters, collection tubes (2mL), Collection tubes (1.5mL), buffers, RNase-free rDNase, carrier RNA, reducing agent TCEP | |
1 Each |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate, ethanol (diluted < 20 %) (denaturedwith 1%IPA/1%MEK, acc.2016/1867/EU); tr